
D(-)-Fructose

Synonym(s):
D(-)-Fructose
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing