
DL-2-Amino-5-phosphonopentanoic acid

Synonym(s):
5-Phosphono-DL-norvaline, DL-2-Amino-5-phosphonovaleric acid, AP-5, APV
Empirical Formula (Hill Notation):
C5H12NO5P
CAS Number:
Molecular Weight:
197.13
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing