
Dimethyl glutarate

Synonym(s):
Dimethyl glutarate, Glutaric acid dimethyl ester
Linear Formula:
CH3OCO(CH2)3COOCH3
CAS Number:
Molecular Weight:
160.17
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing