
Dibutyl phthalate-3,4,5,6-d4

Synonym(s):
Di-n-butyl phthalate-d4
Linear Formula:
C6D4-1,2-[CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
282.37
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing