
Dansylcadaverine

Synonym(s):
N-(5-Aminopentyl)-5-dimethylaminonaphthalene-1-sulfonamide, N-(Dimethylaminonaphthalenesulfonyl)-1,5-pentanediamine, Monodansyl cadaverine
Empirical Formula (Hill Notation):
C17H25N3O2S
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing