
Cytidine 5′-triphosphate disodium salt hydrate

Synonym(s):
5-CTP-Na2, CTP
Empirical Formula (Hill Notation):
C9H14N3Na2O14P3 · xH2O
CAS Number:
Molecular Weight:
527.12 (anhydrous basis)
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing