
Corynoline

Synonym(s):
(+)-Corynoline, 13-Methylchelidonine
Empirical Formula (Hill Notation):
C21H21NO5
CAS Number:
Molecular Weight:
367.40
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing