
Cordycepin

Synonym(s):
3′-Deoxyadenosine
Empirical Formula (Hill Notation):
C10H13N5O3
CAS Number:
Molecular Weight:
251.24
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing