
cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester

Synonym(s):
Methyl 4,7,10,13,16,19-docosahexaenoate
Linear Formula:
CH3(CH2CH=CH)6CH2CH2CO2CH3
CAS Number:
Molecular Weight:
342.51
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing