
cis-4,7,10,13,16,19-Docosahexaenoic acid ethyl ester

Empirical Formula (Hill Notation):
C24H36O2
CAS Number:
Molecular Weight:
356.54
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing