
Cilostazol

Synonym(s):
6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)-butoxy]-3,4-dihydro-2(1H)-quinolinone, OPC 13013, OPC 21, Pletaal, Cilostazol
Empirical Formula (Hill Notation):
C20H27N5O2
CAS Number:
Molecular Weight:
369.46
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-3 of 3