
Chondroitin sulfate A sodium salt from bovine trachea

Synonym(s):
Alternating Copoly β-glucuronic acid-(1→3)-N-acetyl-β-galactosamine-4-sulfate-(1→4)
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Product Line
Tissue
Quality Segment
Package Size
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing