
Cholesterol

Synonym(s):
3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol, Cholesterol
Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Tissue
Quality Segment
Geographic Origin
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing