
Carnosol

Synonym(s):
Picrosalvin
Empirical Formula (Hill Notation):
C20H26O4
CAS Number:
Molecular Weight:
330.42
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Tissue
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing