
Carboxypeptidase Y from baker’s yeast (S. cerevisiae)

Synonym(s):
Peptidyl-L-amino acid Hydrolase, Serine Carboxypeptidase
CAS Number:
Products
Product Category
Brand
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing