
Calcium ascorbate dihydrate

Synonym(s):
Calcium L-ascorbate dihydrate, L-Ascorbic acid calcium salt dihydrate, Vitamin C calcium salt
Empirical Formula (Hill Notation):
C12H14CaO12 · 2H2O
CAS Number:
Molecular Weight:
426.34
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing