
Caffeine Impurity C

Synonym(s):
1,3,9-Trimethylxanthine, 2,6-Dihydroxy-1,3,9-trimethylpurine, Isocaffeine
Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
Products
Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing