
Butylboronic acid N,N,N′,N′-tetramethyl-L-tartaric acid diamide ester

Synonym(s):
(4R,5R)-2-Butyl-N,N,N′,N′-tetramethyl-1,3,2-dioxaborolane-4,5-dicarboxamide
Empirical Formula (Hill Notation):
C12H23BN2O4
CAS Number:
Molecular Weight:
270.13
Products
Boiling Point (°C)
Formula Weight
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing