
Brimonidine tartrate

Synonym(s):
Brimonidine tartrate
Empirical Formula (Hill Notation):
C15H16BrN5O6
CAS Number:
Molecular Weight:
442.22
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-3 of 3