
Bisacodyl Related Compound A

Synonym(s):
4,4′-(2-Pyridinylmethylene)bisphenol, 4,4′-(Pyridin-2-ylmethylene)diphenol, DDPM, Deacetylbisacodyl
Empirical Formula (Hill Notation):
C18H15NO2
CAS Number:
Molecular Weight:
277.32
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing