
Bis-(1,7-dihydro-6H-purine)-6,6,-disulfide

Synonym(s):
6,6′-Dithiodi-9H-purine
Empirical Formula (Hill Notation):
C10H6N8S2
CAS Number:
Molecular Weight:
302.34
Products
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing