Skip to Content
BA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benzenesulfonic acid
Synonym(s):
Benzenesulfonic acid
Linear Formula:
C
6
H
5
SO
3
H
CAS Number:
98-11-3
Molecular Weight:
158.18
Products
Product Category
organic acids (2)
acids (1)
solvents (1)
viscosity reducing agents (1)
Brand
SAFC (1)
Sigma-Aldrich (3)
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technical (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
12635
Benzenesulfonic acid,
98.0% (T)
Expand
View Pricing
1.03893
Benzenesulfonic acid,
EMPROVE
®
EXPERT
Expand
View Pricing
8.00468
Benzenesulfonic acid,
for synthesis
Expand
View Pricing
135070
Benzenesulfonic acid,
technical grade, 90%
Expand
View Pricing