
Benzathine benzylpenicillin for peak identification

Synonym(s):
Benzathine benzylpenicillin
Empirical Formula (Hill Notation):
C32H36N4O8S2 · C16H20N2
CAS Number:
Molecular Weight:
909.12
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing