
Azithromycin for system suitability

Synonym(s):
Azithromycin dihydrate, 1-Oxa-6-azacyclopentadecan-15-one, 13-[(2,6-dideoxy-3-C-methyl-3-O-methyl-alpha-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10- trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-beta-D-xylohexopyranosyl]oxy]-, [2R-(2R, 3S, 4R, 5R, 8R, 10R, 11R, 12S, 13S, 14R)]- dihydrate, N-Methyl-11-aza-10-deoxo-10-dihydroerythromycin A, CP-62993
Empirical Formula (Hill Notation):
C38H72N2O12 · 2H2O
CAS Number:
Molecular Weight:
785.02
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing