
Atorvastatin Related Compound H

Synonym(s):
ATORVASTATIN RELATED COMPOUND H, (Lactone Impurity, 5-(4-Fluorophenyl)-1-{2-[(2R,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-
yl]ethyl}-2-isopropyl-N,4-diphenyl-1H-pyrrole-3-carboxamide)
yl]ethyl}-2-isopropyl-N,4-diphenyl-1H-pyrrole-3-carboxamide)
Empirical Formula (Hill Notation):
C33H33FN2O4
CAS Number:
Molecular Weight:
540.62
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2