
Ascorbic Acid Impurity G

Synonym(s):
(2R)-2-[(2R)-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl]-2-hydroxyacetic acid
Empirical Formula (Hill Notation):
C6H6O7
Molecular Weight:
190.11
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing