
Anti-Dog IgG (whole molecule) antibody produced in rabbit

Products
Product Category
Brand
Biological Source
Clonality
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing