
Anti-Cytokeratin, pan antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-pan-Cytokeratin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2