Skip to Content
BA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Alumina/Silica
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
GF31811060
Alumina/Silica,
fiber, Al2O3 80%/SiO2 20%, tex number 205, length 1 m, filament diameter 0.01mm
Expand
View Pricing
GF65194050
Alumina/Silica,
fiber, Al2O3 80%/SiO2 20%, tex number 205, length 100 m, filament diameter 0.01mm
Expand
View Pricing
GF31721554
Alumina/Silica,
fiber, Al2O3 80%/SiO2 20%, tex number 205, length 50 m, filament diameter 0.01mm
Expand
View Pricing
GF93853772
Alumina/Silica,
fiber, Al2O3 80%/SiO2 20%, tex number 205, length 20 m, filament diameter 0.01mm
Expand
View Pricing
GF76383333
Alumina/Silica,
fiber, tex number 205, Al2O3 80%/SiO2 20%, length 2 m, filament diameter 0.01mm
Expand
View Pricing
GF89550838
Alumina/Silica,
fiber, tex number 205, Al2O3 80%/SiO2 20%, length 10 m, filament diameter 0.01mm
Expand
View Pricing
GF47916401
Alumina/Silica,
fiber, Al2O3 80%/SiO2 20%, tex number 205, length 5 m, filament diameter 0.01mm
Expand
View Pricing