
Ajmalicine

Synonym(s):
δ-Yohimbine, Py-Tetrahydroserpentine, Raubasine
Empirical Formula (Hill Notation):
C21H24N2O3
CAS Number:
Molecular Weight:
352.43
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing