
Adrenalone hydrochloride

Synonym(s):
3′,4′-Dihydroxy-2-(methylamino)acetophenone hydrochloride, Adrenalone hydrochloride
Empirical Formula (Hill Notation):
C9H11NO3 · HCl
CAS Number:
Molecular Weight:
217.65
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2