
Adrenaline tartrate with impurity A

Synonym(s):
(−)-Epinephrine (+)-bitartrate salt, L-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol D-hydrogen bitartrate salt, L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate, L-Epinephrine D-hydrogentartrate
Empirical Formula (Hill Notation):
C9H13NO3 · C4H6O6
CAS Number:
Molecular Weight:
333.29
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing