
6-Methyl-5-hepten-2-ol

Linear Formula:
(CH3)2C=CHCH2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
128.21
Products
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing