
5-(N,N-Dimethyl)amiloride hydrochloride

Synonym(s):
3-Amino-N-(aminoiminoethyl)-5-(dimethylamino)-6-chloropyrazinecarboxamide hydrochloride, DMA
Empirical Formula (Hill Notation):
C8H12ClN7O · HCl
CAS Number:
Molecular Weight:
294.14
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing