
4-tert-Octylphenol monoethoxylate solution

Synonym(s):
2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethanol
Empirical Formula (Hill Notation):
C16H26O2
CAS Number:
Molecular Weight:
250.38
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing