
4-Hydroxy-3-methoxyphenylglycol sulfate potassium salt

Synonym(s):
(3-Methoxy-4-sulfonyloxyphenyl)glycol potassium salt, MHPG sulfate potassium salt, MOPEG sulfate potassium salt
Empirical Formula (Hill Notation):
C9H11KO7S
CAS Number:
Molecular Weight:
302.34
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing