Skip to Content
Merck
4-Heptanone analytical standard

4-Heptanone

4-Heptanone analytical standard
Synonym(s):
Butyrone, Dipropyl ketone
Linear Formula:
(CH3CH2CH2)2CO
CAS Number:
Molecular Weight:
114.19
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing