
4-Amino-3-methylphenol

Synonym(s):
2-Amino-5-hydroxytoluene, 4-Amino-m-cresol, 4-Hydroxy-2-methylaniline
Linear Formula:
H2NC6H3(CH3)OH
CAS Number:
Molecular Weight:
123.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing