
3-sn-Lysophosphatidylethanolamine from egg yolk

Synonym(s):
1-Acyl-sn-glycero-3-phospho(2-aminoethanol), L-α-Lysocephalin, L-α-Lysophosphatidylethanolamine
Products
Product Category
Brand
Physical Form
Purity
Tissue
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing