
3-Methyl-2-oxazolidinone

Synonym(s):
3-Methyl-1,3-oxazolidin-2-one
Empirical Formula (Hill Notation):
C4H7NO2
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing