![3-Fluoro-propylamine hydrochloride AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/306/201/44672225-e56f-41e1-9ddf-ce020bda9406/640/44672225-e56f-41e1-9ddf-ce020bda9406.png)
3-Fluoro-propylamine hydrochloride
![3-Fluoro-propylamine hydrochloride AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/306/201/44672225-e56f-41e1-9ddf-ce020bda9406/640/44672225-e56f-41e1-9ddf-ce020bda9406.png)
Empirical Formula (Hill Notation):
C3H9ClFN
CAS Number:
Molecular Weight:
113.56
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing