
3,7-Dimethyl-1-propargylxanthine

Synonym(s):
3,7-Dimethyl-1-(2-propynyl)xanthine, DMPX
Empirical Formula (Hill Notation):
C10H10N4O2
CAS Number:
Molecular Weight:
218.21
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing