
3,3′-5,5′-Tetraisopropyldiphenol

Synonym(s):
Dipropofol
Empirical Formula (Hill Notation):
C24H34O2
CAS Number:
Molecular Weight:
354.53
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing