Skip to Content
Merck
2-Pentenoic acid predominantly trans, 98%

2-Pentenoic acid

2-Pentenoic acid predominantly trans, 98%
Synonym(s):
trans-2-Pentenoic acid
Linear Formula:
C2H5CH=CHCO2H
CAS Number:
Molecular Weight:
100.12
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing