Skip to Content
Merck
2-Methylindene 98%

2-Methylindene

2-Methylindene 98%
Empirical Formula (Hill Notation):
C10H10
CAS Number:
Molecular Weight:
130.19

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing