
2-Methoxyphenylmagnesium bromide solution

Linear Formula:
CH3OC6H4MgBr
CAS Number:
Molecular Weight:
211.34
Products
Brand
Boiling Point (°C)
Formula Weight
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing