Skip to Content
Merck
2-Chloropyridine-3-carbonyl chloride 98%

2-Chloropyridine-3-carbonyl chloride

2-Chloropyridine-3-carbonyl chloride 98%
Synonym(s):
2-Chloronicotinoyl chloride
Empirical Formula (Hill Notation):
C6H3Cl2NO
CAS Number:
Molecular Weight:
176.00

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing