Skip to Content
Merck
2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein tetrakis(acetoxymethyl) ester BioReagent, suitable for fluorescence

2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein tetrakis(acetoxymethyl) ester

2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein tetrakis(acetoxymethyl) ester BioReagent, suitable for fluorescence
Synonym(s):
BCECF-AM
Empirical Formula (Hill Notation):
C39H36O19
Molecular Weight:
808.69

Products

Brand
Formula Weight
Physical Form
Product Line