Skip to Content
Merck
2,6-Dimethyl-4-heptanone ≥99%

2,6-Dimethyl-4-heptanone

2,6-Dimethyl-4-heptanone ≥99%
Synonym(s):
Diisobutyl ketone, Isovalerone
Linear Formula:
(CH3)2CHCH2COCH2CH(CH3)2
CAS Number:
Molecular Weight:
142.24
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Chemical Composition
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing