2-(4-tert-Butyl-3-hydroxy-2,6-dimethylphenyl)acetamide
Synonym(s):
2-[4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide
Empirical Formula (Hill Notation):
C14H21NO2
CAS Number:
Molecular Weight:
235.32
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Special Grade
Showing 1-1 of 1